missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Granzyme G Polyclonal Antibody
GREENER_CHOICE

Product Code. 15597612
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15597612 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15597612 Supplier Invitrogen™ Supplier No. PA547125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

In Western blots, approximately 5% cross-reactivity with recombinant mouse Granzyme D is observed and less than 1% cross-reactivity with recombinant human Granzyme H is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Granzyme G
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Gzmg
Gene Accession No. P13366
Gene Alias AKA granzyme G; CTL serine protease 1; CTL serine protease 3; Ctla7; Ctla-7; granzyme G; GrzG; Gzmg; MCSP-1
Gene Symbols Gzmg
Host Species Goat
Immunogen Sf 21-derived recombinant mouse Granzyme G Ile21-Leu248.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14944
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.