missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme K Antibody (2471A), DyLight 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB10216K
This item is not returnable.
View return policy
Description
Granzyme K Monoclonal antibody specifically detects Granzyme K in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
Specifications
| Granzyme K | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| 2471A | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, Fragmentin-3, granzyme 3, granzyme K, granzyme K (granzyme 3, tryptase II), granzyme K (serine protease, granzyme 3, tryptase II), Granzyme-3, NK-Tryp-2, NK-tryptase-2, PRSS, TRYP2granzyme-3, tryptase II | |
| Synthetic peptide containing human Granzyme K, Accession # P49863 | |
| 0.1 mL | |
| Apoptosis, Immunology | |
| 3003 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction