missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRAP2 Antibody (OTI1G2), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71457PEATT594
This item is not returnable.
View return policy
Description
GRAP2 Monoclonal specifically detects GRAP2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| GRAP2 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| GRAP2 | |
| Full length recombinant protein of human GRAP2 (NP_004801) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI1G2 | |
| Flow Cytometry | |
| Adapter protein GRID, GADSSH3-SH2-SH3 adapter Mona, GRB2L, GRB-2-like protein, GRB2-related adapter protein 2, GRB2-related adaptor protein 2, GRB2-related protein with insert domain, GRBLGGRAP-2, GRBX, grf-40, Grf40, Grf40 adapter protein, GRIDGRPL, Growth factor receptor-binding protein, growth factor receptor-bound protein 2-related adaptor protein 2, Hematopoietic cell-associated adapter protein GrpL, hematopoietic cell-associated adaptor protein GRPL, Mona, P38, Protein GADS, SH3-SH2-SH3 adaptor molecule | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 9402.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction