Learn More
Invitrogen™ GRAP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597154
Description
Immunogen sequence: MEAVAKFDFT ASGEDELSFH TGDVLKILSN QEEWFKAELG SQEGYVPKNF IDIQFPKWFH EGLSRHQAEN LLMGKEVGFF IIRASQSSPG DFSISVRHED DVQHFKVMRD NKGNYFLWTE KFPSLNKLVD YYRTNSISRQ KQIFLRDRTR EDQGHRGNSL DRRSQGGPHL SGAVGEEIRP SMNRKLSDHP PTLPLQQHQH QPQPPQYAPA PQQLQQPPQQ RYLQHHHFHQ ERRGGSLDIN DGHCGTGLGS EMNAALMHRR HTDPVQLQAA GRVRWARALY DFEALEDDEL GFHSGEVVEV LDSSNPSWWT GRLHNKLGLF PANYVAPMTR; Positive Samples: Jurkat; Cellular Location: Cytoplasm, Endosome, Nucleus.
This gene encodes a member of the GRB2/Sem5/Drk family. This member is an adaptor-like protein involved in leukocyte-specific protein-tyrosine kinase signaling. Like its related family member, GRB2-related adaptor protein (GRAP), this protein contains an SH2 domain flanked by two SH3 domains. This protein interacts with other proteins, such as GRB2-associated binding protein 1 (GAB1) and the SLP-76 leukocyte protein (LCP2), through its SH3 domains. Transcript variants utilizing alternative polyA sites exist.
Specifications
| GRAP2 | |
| Polyclonal | |
| Unconjugated | |
| Grap2 | |
| adapter protein GRID; Gads; GADS protein; Grap2; GRAP-2; GRB2L; GRB-2-like protein; GRB2-related adapter protein 2; Grb2-related adaptor downstream of Sch; GRB2-related adaptor protein 2; GRB-2-related monocytic adapter protein; GRB2-related protein with insert domain; GRBLG; GRBX; Grf40; grf-40; Grf40 adapter protein; GRID; Growth factor receptor-binding protein; growth factor receptor-bound protein 2-related adaptor protein 2; GrpL; Hematopoietic cell-associated adapter protein GrpL; hematopoietic cell-associated adaptor protein GRPL; MONA; monocytic adapter; monocytic adaptor; P38; Protein GADS; RP3-370M22.1; SH3-SH2-SH3 adapter Mona; SH3-SH2-SH3 adaptor molecule | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17444, 9402 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.76 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75791, O89100 | |
| Grap2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-330 of human GRAP2 (NP_0048011). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.