Learn More
Trajan Scientific and Medical™ Short Reducing Ferrules for 1/16 in. Fittings
15% Graphite / 85% Vespel ferrules provide leak-free system connections, leading to improved chromatography for non-MS systems
Brand:  Trajan Scientific and Medical™ 073111
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11902637
  • € 65.75 / Pack of 10
Estimated Shipment: 28-07-2021
Add to basket
Description
Description
  • Composite of graphite and Vespel
  • Mechanically robust, hard material, long lifetime
  • Forms a strong grip with the capillary column
Specifications
Specifications
2.7 mm
1/16 in. Fittings
Reducing Ferrule
0.32 mm
3.2 mm
Graphite/Vespel
10/Pk.