missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRASP1 Antibody (OTI1E3), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72064IR
This item is not returnable.
View return policy
Description
GRASP1 Monoclonal specifically detects GRASP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| GRASP1 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GRIPAP1 | |
| Full length recombinant protein of human GRASP1 (NP_064522) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1E3 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DKFZp434P0630, GRASP1, GRASP-1MGC126595, GRIP1 associated protein 1, GRIP1-associated protein 1, KIAA1167MGC126593, MPMGp800B12492Q3 | |
| Mouse | |
| Affinity Purified | |
| Neurotransmission | |
| 56850.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction