missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRASP55 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99119-100ul
This item is not returnable.
View return policy
Description
GRASP55 Polyclonal antibody specifically detects GRASP55 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GRASP55 | |
| Polyclonal | |
| Immunohistochemistry 1:500-1:2000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Golgi phosphoprotein 6, golgi reassembly stacking protein 2, 55 kDa, golgi reassembly stacking protein 2, 55kDa, Golgi reassembly-stacking protein of 55 kDa, GOLPH6DKFZp434D156, GRASP55FLJ13139, GRS2Golgi reassembly-stacking protein 2, p59 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human GRASP55. | |
| 100 μg | |
| Golgi Apparatus Markers | |
| 26003 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction