missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRASP55 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581185
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human GORASP2.
GRASP55 is a PDZ containing protein involved in the stacking of golgi cisternea in a cell-free system.
Specifications
| GRASP55 | |
| Polyclonal | |
| Unconjugated | |
| GORASP2 | |
| 0610011A07Rik; 2410043M02Rik; 5730520M13Rik; 9430094F20Rik; AW552058; golgi phosphoprotein 6; golgi reassembly stacking protein 2; golgi reassembly stacking protein 2, 55kDa; Golgi reassembly-stacking protein 2; Golgi reassembly-stacking protein of 55 kDa; GOLPH2; GOLPH6; Gorasp2; GRASP55; GRS2; LOW QUALITY PROTEIN: Golgi reassembly-stacking protein 2; p59 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 26003 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9H8Y8 | |
| GORASP2 | |
| A synthetic peptide corresponding to the N-terminus of the Human GORASP2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction