missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRASP65 Antibody (OTI5G8), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72144AF532
This item is not returnable.
View return policy
Description
GRASP65 Monoclonal specifically detects GRASP65 in Human, Rat, Porcine, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| GRASP65 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| GORASP1 | |
| Full length recombinant protein of human GRASP65 (NP_114105) produced in HEK293T cells. | |
| Primary | |
| Human, Rat, Pig, Canine, Primate, Monkey | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI5G8 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Golgi peripheral membrane protein p65, Golgi phosphoprotein 5MGC118897, golgi reassembly stacking protein 1, 65kDa, Golgi reassembly-stacking protein 1, Golgi reassembly-stacking protein of 65 kDa, GRASP65MGC118894, P6565 kDa | |
| Mouse | |
| 0.1 mL | |
| 64689 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction