missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRASP65 Monoclonal Antibody (OTI5G8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525148
This item is not returnable.
View return policy
Description
GRASP65 Monoclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot
Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.
Specifications
| GRASP65 | |
| Monoclonal | |
| 0.6 mg/mL | |
| PBS with 50% glycerol, 1% BSA and 0.02% sodium azide; pH 7.3 | |
| O35254, Q9BQQ3 | |
| GORASP1 | |
| Full length human recombinant protein of GORASP1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Rat, Canine, Monkey | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI5G8 | |
| Unconjugated | |
| GORASP1 | |
| 5430411C10Rik; Golgi peripheral membrane protein p65; Golgi phosphoprotein 5; Golgi reassembly and stacking protein 1; golgi reassembly stacking protein 1; golgi reassembly stacking protein 1, 65kDa; Golgi reassembly-stacking protein 1; Golgi reassembly-stacking protein of 65 kDa; GOLPH5; Gorasp1; GRASP65; P65 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 485594, 56082, 64689 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction