missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRB 14 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97218PE
This item is not returnable.
View return policy
Description
GRB 14 Polyclonal antibody specifically detects GRB 14 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| GRB 14 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| GRB14 adapter protein, growth factor receptor-bound protein 14 | |
| Produced in rabbits immunized with E. coli-derived Human GRB 14 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 2888 | |
| Store at 4C in the dark. | |
| IgG |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction