missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRB10 Monoclonal Antibody (OTI5A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF812343
This item is not returnable.
View return policy
Description
GRB10 Monoclonal antibody specifically detects GRB10 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| GRB10 | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O43665, P49806, Q9CQE5 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 54290, 6001, 67865 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5A6 | |
| Unconjugated | |
| RGS10 | |
| 5730571D09Rik; AI325020; Grb10; Grb-10; GRB10 adapter protein; GRB10 adaptor protein; GRBIR; GRB-IR; growth factor receptor bound protein 10; growth factor receptor-bound protein 10; Insulin receptor-binding protein Grb-IR; IRBP; KIAA0207; maternally expressed gene 1; maternally expressed gene 1 protein; Meg1; mKIAA0207; OTTHUMP00000197988; OTTHUMP00000197989; OTTHUMP00000197990; OTTHUMP00000197991; OTTHUMP00000197993; OTTHUMP00000197994; RGD1566234; RSS | |
| Full length human recombinant protein of human RGS10 (NP_001005339) produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction