missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRB10 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 81225-1-RR-100UL
This item is not returnable.
View return policy
Description
The GRB10 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human GRB10. This antibody recognizes human, mouse, rat antigen. The GRB10 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| GRB10 | |
| Recombinant Monoclonal | |
| 1000 μg/mL | |
| GRB-IR; Grb-10; IRBP; KIAA0207; MEG1; RSS | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 60-70 kDa | |
| Liquid |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| 1D13 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 2887 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction