missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GRB2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA527151

Product Code. 13481217

  • € 586.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: 293T, A431, HeLa, HepG2, NIH-3T3, BCL-1. Predicted reactivity: Mouse (100%), Rat (100%), Pig (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GRB 2 protein contains one SH2 domain flanked on both sides by an SH3 domain. In response to EGF, GRB 2 binds, through its SH3 domains, to the guanine nucleotide exchange factor, Sos. The GRB 2-Sos complex then binds the EGF receptor. These interactions play a critical role in the activation of the p21 ras signaling pathway.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GRB2
Polyclonal
Unconjugated
GRB2
AA408164; abundant SRC homology; adapter protein GRB2; Adapter protein GRB2-A; ASH; ASH protein; EGFRBP-GRB2; epidermal growth factor receptor-binding protein GRB2; grb2; grb2.L; grb2-a; grb2-b; Grb3-3; growth factor receptor bound protein 2; growth factor receptor bound protein 2 L homeolog; growth factor receptor-bound protein 2; Growth factor receptor-bound protein 2-A; growth factor receptor-bound protein 3; HT027; hypothetical protein LOC535298; MST084; MSTP084; NCKAP2; OTTHUMP00000166097; OTTHUMP00000166098; protein Ash; SH2/SH3 adapter GRB2; SH2/SH3 adapter GRB2-A; SH2/SH3 adaptor Grb2; XELAEV_18044055mg
Rabbit
Antigen affinity chromatography
RUO
14784, 2885, 81504
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
P62993, P62994, Q60631
GRB2
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human GRB2. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.