missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | GRB2 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16856523
|
Proteintech
10254-2-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16846523
|
Proteintech
10254-2-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRB 2 protein contains one SH2 domain flanked on both sides by an SH3 domain. In response to EGF, GRB 2 binds, through its SH3 domains, to the guanine nucleotide exchange factor, Sos. The GRB 2 - Sos complex then binds the EGF receptor. These interactions play a critical role in the activation of the p21 ras signaling pathway.Specifications
| GRB2 | |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| P62993, P62994, Q60631 | |
| 14784, 2885, 81504 | |
| GRB2 Fusion Protein Ag0395 | |
| Primary | |
| -20°C | |
| GRB2 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Adapter protein GRB2, ASH, EGFRBP GRB2, GRB2, Grb3 3, MST084, MSTP084, Protein Ash, SH2/SH3 adapter GRB2 | |
| GRB2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title