missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRB7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144627
This item is not returnable.
View return policy
Description
Positive control: SK-Br-3 cell lysates. Predicted band size: 60 kDa Subcellular Location: Cell membrane, Cytoplasm, Focal adhesion, Cytoplasmic granule, Cell projection.
The product of this gene belongs to a small family of adapter proteins that are known to interact with a number of receptor tyrosine kinases and signaling molecules. This gene encodes a growth factor receptor-binding protein that interacts with epidermal growth factor receptor (EGFR) and ephrin receptors. The protein plays a role in the integrin signaling pathway and cell migration by binding with focal adhesion kinase (FAK). Alternative splicing results in multiple transcript variants encoding different isoforms, although the full-length natures of only two of the variants have been determined to date.
Specifications
| GRB7 | |
| Polyclonal | |
| Unconjugated | |
| GRB7 | |
| B47; Epidermal growth factor receptor GRB-7; Grb7; GRB7 adapter protein; growth factor receptor binding protein GRB7; growth factor receptor bound protein 7; growth factor receptor-bound protein 7; Growth factor receptor-bound protein 7 (GRB7 adapter protein) (Epidermal growth factor receptor GRB-7); mKIAA4028; OTTHUMP00000164352 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2886 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS, TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q14451 | |
| GRB7 | |
| Recombinant protein within human GRB7 aa 1-200/532. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction