missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRCC10 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97383V
This item is not returnable.
View return policy
Description
GRCC10 Polyclonal antibody specifically detects GRCC10 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GRCC10 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 113246 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| C10, C10GRCC10, chromosome 12 open reading frame 57, GRCC10, likely ortholog of mouse gene rich cluster, C10, protein C10 | |
| Produced in rabbits immunized with E. coli-derived Human GRCC10 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction