Learn More
GREB1 Antibody (CL13859) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24583IR
Product Code. 30291052
  • € 737.00 / 0.10mL
Description
GREB1 Monoclonal antibody specifically detects GREB1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GREB1
Monoclonal
DyLight 755
gene regulated by estrogen in breast cancer protein, growth regulation by estrogen in breast cancer 1, KIAA0575Gene regulated in breast cancer 1 protein, protein GREB1
This antibody was developed using a recombinant protein derived from Q4ZG55, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL13859
50mM Sodium Borate
Mouse
Protein A purified
RUO
9687
Store at 4⪚C in the dark.
IgG2a