missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GREB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115324
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GREB1.
This gene is an estrogen-responsive gene that is an early response gene in the estrogen receptor-regulated pathway. It is thought to play an important role in hormone-responsive tissues and cancer. Three alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| GREB1 | |
| Polyclonal | |
| Unconjugated | |
| GREB1 | |
| 5730583K22Rik; 9130004E13; AF180470; AU023194; gene regulated by estrogen in breast cancer; gene regulated by estrogen in breast cancer protein; gene regulated in breast cancer 1 protein; GREB1; growth regulating estrogen receptor binding 1; growth regulating estrogen receptor binding 1; protein GREB1; growth regulation by estrogen in breast cancer 1; id:ibd5087; id:ibd5087 protein; im:7140582; Kiaa0575; mKIAA0575; protein GREB1; protein GREB1-like; RGD1564091; si:ch211-175f11.3; zgc:158681 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 268527, 500633, 9687 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q3UHK3, Q4ZG55 | |
| GREB1 | |
| A synthesized peptide derived from human GREB1(Accession Q4ZG55), corresponding to amino acid residues S1300-G1350. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction