Learn More
Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes
Product Code. 13058592
Change view
Click to view available options
Capacity (Metric):
0.8 mL
1 mL
Additive:
K3 EDTA
Serum Sep
Sodium Citrate (0.105M)
Closure Color:
Blue
Gold
Lavender
Unit Size:
Pack of 100
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Additive Closure Color Unit Size
13058592 1 mL Sodium Citrate (0.105M) Blue Pack of 1000
12671501 0.8 mL Serum Sep Gold Pack of 100
12500127 1 mL K3 EDTA Lavender Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13058592 Supplier Greiner Bio-One Supplier No. 450413
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Collect, transport and process capillary blood. Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes are especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required.
  • Tubes feature closure that acts as a valve, allowing appropriate capillary or funnel to be inserted without removing the cap
  • Fit into a 13 × 75mm carrier tube to facilitate bar code labeling, centrifugation, and storage
  • Tubes with gel contain an inert barrier on the bottom that moves upward to form a stable barrier between the clot/cells and serum/plasma during centrifugation and transport
  • All tubes require MiniCollect funnel or capillary collection device (sold separately)
TRUSTED_SUSTAINABILITY
Specifications
Product Type MiniCollect Capillary Blood Collection Tube
Additive Sodium Citrate (0.105M)
Capacity (Metric) 1 mL
Closure Included Yes
Closure Color Blue
For Use With (Application) Small volume blood collection