Learn More
Greiner Bio-One Speedy Quick Release Tube Holder
Product Code. 10748315
Change view
Click to view available options
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
10748315 Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10748315 Supplier Greiner Bio-One Supplier No. 450212
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Trigger mechanism for safe and easy needle ejection
Speedy Quick Release Tube Holder
  • After venipuncture the needle is simply released from the holder by pushing the release mechanism so that the needle falls directly into a sharps disposal container
  • The release mechanism is designed to retract automatically
  • It is recommended to use this device as a single-use holder
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tube Holder