missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRF2 Antibody (OTI2F5), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70846UV
This item is not returnable.
View return policy
Description
GRF2 Monoclonal specifically detects GRF2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| GRF2 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RAPGEF1 | |
| Full length recombinant protein of human GRF2 (NP_941372) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2F5 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| C3GGRF2guanine nucleotide-releasing factor 2 (specific for crk proto-oncogene), CRK SH3-binding GNRP, DKFZp781P1719, Guanine nucleotide-releasing factor 2, Protein C3G, Rap guanine nucleotide exchange factor (GEF) 1, rap guanine nucleotide exchange factor 1 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 2889.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction