missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRG (Groucho homolog) Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 369.00 - € 593.00
Specifications
| Antigen | GRG (Groucho homolog) |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18295643
|
Novus Biologicals
NBP2-58543 |
100 μL |
€ 593.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18662527
|
Novus Biologicals
NBP2-58543-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRG (Groucho homolog) Polyclonal specifically detects GRG (Groucho homolog) in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| GRG (Groucho homolog) | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Signal Transduction, Wnt Signaling Pathway | |
| AES-1, AES-2, Amino enhancer of split, amino-terminal enhancer of split, ESP1, Gp130-associated protein GAM, GRG, GRG5, Protein ESP1, Protein GRG, TLE5 | |
| TLE5 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 166 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MCHKNGFPQEGGITAAFLQKRKLRLSKNHRPARAKVTEHVRGTRPGRATAGPAASTRAAGSLFFDRWGNRGPAGCRGSSHLPQQLKFTTSD | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title