missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRHL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552210
This item is not returnable.
View return policy
Description
Immunogen sequence: TVFKPFIDLD TQPVLFIPDV HFANLQRGTH VLPIASEELE GEGSVLKRGP YGTEDDFAVP PSTKLARIEE PKRVLLYVRK ESEEVFDALM LKTPSLKGL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.
This gene encodes a member of the grainyhead family of transcription factors. The encoded protein can exist as a homodimer or can form heterodimers with sister-of-mammalian grainyhead or brother-of-mammalian grainyhead. This protein functions as a transcription factor during development.
Specifications
| GRHL1 | |
| Polyclonal | |
| Unconjugated | |
| GRHL1 | |
| DNA methyltransferase 3A; Dnmt3a; grainyhead like 1; grainyhead like transcription factor 1; grainyhead-like 1; grainyhead-like 1 (Drosophila); grainyhead-like protein 1 homolog; grainyhead-like transcription factor 1; GRHL1; LBP protein 32; LBP32; LBP-32; Mammalian grainyhead; Mgr; NH32; p61 MGR; p70 MGR; Tcfcp2l2; TFCP2L2; transcription factor CP2-like 2; Transcription factor LBP-32 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 195733, 29841, 313993 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q921D9, Q9NZI5 | |
| GRHL1 | |
| Recombinant protein corresponding to Human GRHL1. Recombinant protein control fragment (Product #RP-89348). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction