missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRHL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598889
This item is not returnable.
View return policy
Description
GRHL1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a member of the grainyhead family of transcription factors. The encoded protein can exist as a homodimer or can form heterodimers with sister-of-mammalian grainyhead or brother-of-mammalian grainyhead. This protein functions as a transcription factor during development.
Specifications
| GRHL1 | |
| Polyclonal | |
| Unconjugated | |
| GRHL1 | |
| DNA methyltransferase 3A; Dnmt3a; grainyhead like 1; grainyhead like transcription factor 1; grainyhead-like 1; grainyhead-like 1 (Drosophila); grainyhead-like protein 1 homolog; grainyhead-like transcription factor 1; GRHL1; LBP protein 32; LBP32; LBP-32; Mammalian grainyhead; Mgr; NH32; p61 MGR; p70 MGR; Tcfcp2l2; TFCP2L2; transcription factor CP2-like 2; Transcription factor LBP-32 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 29841 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3.95 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9NZI5 | |
| GRHL1 | |
| Recombinant Human Grainyhead-like protein 1 homologprotein (1-95AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction