missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRHL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569031
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
The protein encoded by this gene is a transcription factor that can act as a homodimer or as a heterodimer with either GRHL1 or GRHL3. Defects in this gene are a cause of non-syndromic sensorineural deafness autosomal dominant type 28 (DFNA28).
Specifications
| GRHL2 | |
| Polyclonal | |
| Unconjugated | |
| GRHL2 | |
| 0610015A08Rik; BOM; Brother of mammalian grainyhead; DFNA28; ECTDS; grainyhead like 2; grainyhead like transcription factor 2; grainyheadlike; grainyhead-like 2; grainyhead-like 2 (Drosophila); grainyhead-like protein 2 homolog; grainyhead-like transcription factor 2; Grhl2; RGD1561191; Tcfcp2l3; TFCP2L3; transcription factor CP2-like 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 299979 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| GRHL2 | |
| synthetic peptide directed towards the N-terminal region of rat GRHL2. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction