missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRHPR Antibody (OTI2H4), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71430IR
This item is not returnable.
View return policy
Description
GRHPR Monoclonal specifically detects GRHPR in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| GRHPR | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GRHPR | |
| Full length recombinant protein of human GRHPR (NP_036335) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI2H4 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| EC 1.1.1.81, GLXREC 1.1.1.79, glycerate-2-dehydrogenase, GLYD, glyoxylate reductase/hydroxypyruvate reductase, PH2 | |
| Mouse | |
| Affinity Purified | |
| metabolism | |
| 9380.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction