missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRHPR Monoclonal Antibody (OTI10A3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502090
This item is not returnable.
View return policy
Description
This gene encodes an enzyme with hydroxypyruvate reductase, glyoxylate reductase, and D-glycerate dehydrogenase enzymatic activities. The enzyme has widespread tissue expression and has a role in metabolism. Type II hyperoxaluria is caused by mutations in this gene.Specifications
| GRHPR | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UBQ7 | |
| GRHPR | |
| Full length human recombit protein of human GRHPR produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI10A3 | |
| Unconjugated | |
| GRHPR | |
| GLXR, GLYD, PH2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9380 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction