missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRIA1 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27885-1-AP-150UL
This item is not returnable.
View return policy
Description
The GRIA1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GRIA1. This antibody recognizes human, mouse, pig, rat antigen. The GRIA1 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| GRIA1 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2890 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, ELISA | |
| 800 μg/mL | |
| GLUH1; GLUR1; GLURA; HBGR1; MGC133252 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| 102 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction