missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grin2a Mouse anti-Rat, HRP, Clone:S327A-38, Abnova™
Mouse monoclonal antibody raised against partial recombinant rat Grin2a.
Brand: Abnova MAB17347.100ug
This item is not returnable.
View return policy
Specifications
| Grin2a | |
| Monoclonal | |
| HRP | |
| Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
| Grin2a | |
| Mouse | |
| 100 μg | |
| Rat | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| S327A-38 | |
| glutamate receptor, ionotropic, N-methyl D-aspartate 2A | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| NMDAR2A, NR2A | |
| Recombinant protein corresponding to amino acids 75-325 at N-terminus of rat Grin2a. | |
| 24409 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction