Learn More
Kinematica™ Stainless Steel Mixing attachment for MB 800 Laboratory Mixer
Product Code. 11989058 Shop All Kinematica Products
Click to view available options
Capacity (Metric):
2000 mL
4000 mL
Unit Size:
Each
Product Code. 11989058
Brand: Kinematica™ 36050050
Only null left
Add to basket
Add to basket
Mixing attachment for MB 800 consists of stainless steel, gasket and blade head as well as lid with opening for material addition.
Specifications
Capacity (Metric) 2000 mL
Blade Material Stainless Steel
For Use With (Equipment) MB 800