missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRINL1A Monoclonal Antibody (2E4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522667
This item is not returnable.
View return policy
Description
Peptide Sequence: GSPISSEERR RRDKQHLDDI TAARLLPLHH MPTQLLSIEE SLALQKQQKQ NYEEMQAKLA AQKLAERLNI KMRSYNPEGE SSGRYREVRD EDDDWSSDEF.
This gene (Gcom1) is part of a complex transcript unit that includes the gene for glutamate receptor, ionotropic, N-methyl D-aspartate-like 1A (GRINL1A). Transcription of this gene occurs at an upstream promoter, with two different groups of alternatively spliced variants: Gup for GRINL1A upstream transcripts and Gcom for GRINL1A combined transcripts. The GRINL1A gene uses a downstream promoter for transcription and also has multiple alternatively spliced variants.
Specifications
| GRINL1A | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P0CAP2, Q6EEV4 | |
| POLR2M | |
| POLR2M (NP_056347.1, 269 a.a. ~ 368 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2E4 | |
| Unconjugated | |
| POLR2M | |
| AA407243; AA408799; AI303746; AW553238; D9Wsu138e; DNA-directed RNA polymerase II subunit GRINL1A; DNA-directed RNA polymerase II subunit GRINL1A, isoforms 4/5; DNA-directed RNA polymerase II subunit M; DNA-directed RNA polymerase II subunit M, isoforms 4/5; gcom; GCOM1; Gcom2; Gdown; Gdown1; Glurr; glutamate receptor, ionotropic, N-methyl D-aspartate-like 1A; glutamate receptor-like protein 1A; Grinl1a; GRINL1A downstream protein 1; GRINL1A downstream protein Gdown4; GRINL1A upstream protein; Gup; mKIAA4185; Myocardial zonula adherens protein; MYOZAP; MYZAP; MYZAP-POLR2M; POLR2M; polymerase (RNA) II (DNA directed) polypeptide M; polymerase (RNA) II subunit M; protein GRINL1A; protein GRINL1A, isoforms 4/5; RNA polymerase II subunit M | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 81488 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction