missing translation for 'onlineSavingsMsg'
Learn More

GRIP1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB120-2842-0.025mg

 View more versions of this product

Product Code. 18527451

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

GRIP1 Polyclonal specifically detects GRIP1 in Human, Hamster samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GRIP1
Polyclonal
Unconjugated
PBS with 0.05% Sodium Azide
GRIP1
Synthetic peptide corresponding to residues P(206) L P D S E E E G H D S(217) C of human GRIP1.
0.025 mg
Cancer, Neuronal Cell Markers, Neurotransmission, Transcription Factors and Regulators
23426
Human
Purified
Immunocytochemistry
1 mg/mL
Immunocytochemistry/Immunofluorescence 1 - 2 μg/mL
glutamate receptor interacting protein 1, glutamate receptor-interacting protein 1, GRIP, GRIP-1
Rabbit
Immunogen affinity purified
RUO
Primary
GRIP1
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only