missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRIP1 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41211DL594
This item is not returnable.
View return policy
Description
GRIP1 Polyclonal antibody specifically detects GRIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GRIP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| glutamate receptor interacting protein 1, glutamate receptor-interacting protein 1, GRIP, GRIP-1 | |
| Antibody was raised against a 17 amino acid peptide near the carboxy terminus of human GRIP1. The immunogen is located within the last 50 amino acids of GRIP1. Amino Acid Squence: QPSHGGNLETREPTNTL | |
| 0.1 mL | |
| Cancer, Neuronal Cell Markers, Neurotransmission, Transcription Factors and Regulators | |
| 23426 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction