missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRK1 Antibody (G8), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-2775MFV610
This item is not returnable.
View return policy
Description
GRK1 Monoclonal antibody specifically detects GRK1 in Human, Mouse, Rat, Bovine, Chicken samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| GRK1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine, Chicken | |
| Purified |
| Western Blot, Immunocytochemistry, Immunoprecipitation | |
| G8 | |
| Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
| G protein-coupled receptor kinase 1rhodopsin kinase, GPRK1, RHOKEC 2.7.11, RKEC 2.7.11.14 | |
| Full length human GRK1. | |
| 0.1 mL | |
| Protein Kinase, Signal Transduction, Vision | |
| 6011 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction