missing translation for 'onlineSavingsMsg'
Learn More

GRK1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

€ 145.00 - € 410.00

Specifications

Antigen GRK1
Concentration 0.33 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16883095
View Documents
Proteintech
24606-1-AP-20UL
20 μL
€ 145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16873095
View Documents
Proteintech
24606-1-AP-150UL
150 μL
€ 410.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The sensation of sight is the result of a cascade of events starting with the interaction of photoactivated rhodopsin with a protein called transducin. Rhodopsin kinase is a G-protein-coupled Ser/Thr kinase, also known as GRK1, which is a key element in the regulation of this cascade. Following phosphorylation by GRK1, arrestin is recruited to phospho-rhodopsin quenching its phototransductive activity by preventing further interaction with transducin. By breaking the cycle of phototransduction, GRK1 plays an important role in the restoration of the system for subsequent visual events.
Specifications

Specifications

GRK1
Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human, Mouse
Q15835, Q9WVL4
24013, 6011
GRK1 Fusion Protein Ag18063
Primary
-20°C
GRK1
0.33 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
GPRK1, GRK1, Rhodopsin kinase, RHOK, RK
GRK1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.