missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRK4 Antibody (OTI4F5), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70845IR
This item is not returnable.
View return policy
Description
GRK4 Monoclonal specifically detects GRK4 in Human samples. It is validated for Western Blot.
Specifications
| GRK4 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GRK4 | |
| Recombinant protein fragment corresponding to amino acids 1-245 of human GRK4 (NP_892027) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI4F5 | |
| Western Blot | |
| EC 2.7.11, EC 2.7.11.16, G protein-coupled receptor kinase 2-like (Drosophila), G protein-coupled receptor kinase 4, G protein-coupled receptor kinase GRK4, GPRK2LIT11, GPRK4GRK4a, G-protein coupled receptor kinase 4, ITI1 | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase | |
| 2868.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction