missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRK6 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32197B
This item is not returnable.
View return policy
Description
GRK6 Polyclonal antibody specifically detects GRK6 in Human, Mouse samples. It is validated for Western Blot
Specifications
| GRK6 | |
| Polyclonal | |
| Western Blot | |
| EC 2.7.11, EC 2.7.11.16, FLJ32135, G protein-coupled receptor kinase 6, GPRK6G protein-coupled receptor kinase GRK6 | |
| The immunogen this antibody was made to, maps to a region between residue 526 to 576 of human G Protein-Coupled Receptor Kinase 6 using the numbering given in entry NP_001004106.1 (GeneID 2870). | |
| 0.1 mL | |
| Protein Kinase | |
| 2870 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction