missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRK6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101113
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GRK6.
GRK6 is a member of the guanine nucleotide-binding protein coupled receptor kinase subfamily of the serine/threonine protein kinase family. GRK6 phosphorylates the activated forms of G protein-coupled receptors to initiate their deactivation. Several transcript variants encoding different isoforms have been described for GRK6.
Specifications
| GRK6 | |
| Polyclonal | |
| Unconjugated | |
| Grk6 | |
| activin receptor IIA; activin receptor type IIA; Activin receptor type-2A; ActrIIa; ACTR-IIA; Acvr2; Acvr2a; EC 2.7.11.16; G protein-coupled receptor kinase 6; g protein-coupled receptor kinase GRK6; Gprk6; G-protein coupled receptor kinase 6; GRK6; kinase GRK6; TactrII | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 105540, 236304, 26385, 2870, 59076 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O70293, P43250, P97711 | |
| Grk6 | |
| A synthesized peptide derived from human GRK6(Accession P43250), corresponding to amino acid residues E2-D52. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction