missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRK6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11439-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. The protein phosphorylates the activated forms of G protein-coupled receptors thus initiating their deactivation. Several transcript variants encoding different isoforms have been described for this gene.Specifications
| GRK6 | |
| Polyclonal | |
| Unconjugated | |
| GRK6 | |
| GPRK6, GRK6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26385, 2870, 59076 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O70293, P43250, P97711 | |
| GRK6 | |
| GRK6 Fusion Protein Ag1992 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction