missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRO2 Antibody (MM0214-11B27), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12227R
This item is not returnable.
View return policy
Description
GRO2 Monoclonal specifically detects GRO2 in Human samples. It is validated for Western Blot.
Specifications
| GRO2 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CXCL2 | |
| Human recombinant GRO2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| MM0214-11B27 | |
| Western Blot | |
| chemokine (C-X-C motif) ligand 2, CINC-2a, GRO2 oncogene, GRO2MIP2, GROb, gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MIP-2a, MIP2AC-X-C motif chemokine 2, SCYB2MIP2-alpha | |
| Mouse | |
| Protein G purified | |
| Signal Transduction | |
| 2920 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction