missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Growth Hormone Antibody Cocktail, Invitrogen™
Antibody Cocktail
Brand: Invitrogen MA511926
This item is not returnable.
View return policy
Description
MA5-11926 targets Growth Hormone Human in IHC (P) and WB applications and shows reactivity with Human samples. The MA5-11926 immunogen is purified recombinant human growth hormone.
Growth Hormone Cocktail antibody specifically detects Growth Hormone in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Specifications
| Growth Hormone | |
| Cocktail | |
| 400 μg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P01241 | |
| Gh1 | |
| Purified recombinant human growth hormone | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| GH01 (#2), GH02 (#7), GH02 (No.7) | |
| Unconjugated | |
| Gh1 | |
| dog growth hormone; GH; Gh1; GHB1; GHB5; GHN; GH-N; Growth hormone; growth hormone 1; growth hormone 12; growth hormone B1; growth hormone B5; growth hormone precursor; HGH; hGH-N; IGHD1B; pituitary growth hormone; RNGHGP; Somatotropin | |
| Mouse | |
| Protein A | |
| RUO | |
| 2688 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction