missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Growth Hormone Antibody (GH/1450), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54356V
This item is not returnable.
View return policy
Description
Growth Hormone Monoclonal antibody specifically detects Growth Hormone in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Growth Hormone | |
| Monoclonal | |
| DyLight 405 | |
| GHIGHD1B, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N | |
| A recombinant fragment (around aa58-187) of human Growth Hormone (GH) protein (exact sequence is proprietary) (Uniprot: P01241) | |
| 100 μg | |
| BiologicAll speciesy Active Proteins | |
| 2688 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| GH/1450 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction