missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Growth Hormone Antibody (GH/8215R) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20868
This item is not returnable.
View return policy
Description
Growth Hormone Monoclonal antibody specifically detects Growth Hormone in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Growth Hormone | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| GHIGHD1B, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N | |
| Recombinant full-length human Growth Hormone protein | |
| 100 μg | |
| Biologically Active Proteins | |
| 2688 | |
| Store at -20°C to -70°C. Avoid freeze/thaw cycles. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| GH/8215R | |
| Unconjugated | |
| 10 mM PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction