missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Growth Hormone Antibody (GH/8215R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 298.00 - € 530.00
Specifications
| Antigen | Growth Hormone |
|---|---|
| Clone | GH/8215R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18694375
|
Novus Biologicals
NBP3-20274-20ug |
20 μg |
€ 298.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613304
|
Novus Biologicals
NBP3-20274-100ug |
100 μg |
€ 530.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Growth Hormone Monoclonal antibody specifically detects Growth Hormone in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Growth Hormone | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins | |
| 10mM PBS with 0.05% BSA | |
| 2688 | |
| IgG κ | |
| Protein A or G purified |
| GH/8215R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| GHIGHD1B, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N | |
| Recombinant full-length human Growth Hormone protein | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel