missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRP78/HSPA5 Antibody (1H11-1H7), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12888F
This item is not returnable.
View return policy
Description
GRP78/HSPA5 Monoclonal specifically detects GRP78/HSPA5 in Human, Mouse, Rat, Bovine, Fungi, Hamster, Monkey, Rabbit, Xenopus samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| GRP78/HSPA5 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| HSPA5 | |
| His-tagged human GRP78 | |
| 0.1 mL | |
| Primary | |
| Detects a protein with a mass of approx. 78kDa corresponding to GRP78 | |
| Store at 4C in the dark. | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence | |
| 1H11-1H7 | |
| Immunocytochemistry/Immunofluorescence | |
| 78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2 | |
| Mouse | |
| Protein G purified | |
| Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
| 3309 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction