missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GRP78/HSPA5 Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
Specifications
Specifications
| Gene ID (Entrez) | 3309 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB300-520 |
| Gene Alias | 78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70 kD protein 5 (glucose-regulated protein, 78 kD), heat shock 70 kDa protein 5 (glucose-regulated protein, 78 kDa), Immunoglobulin heavy chain-binding protein, MIF2 |
| Gene Symbol | HSPA5 |
| Quantity | 0.1 mg |
| Research Category | Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR) |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction