missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRP78/HSPA5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 209.00
Specifications
| Antigen | GRP78/HSPA5 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000-1:2000, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
GRP78/HSPA5 Polyclonal specifically detects GRP78/HSPA5 in Human, Mouse, Rat, Canine, Drosophila, Hamster samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| GRP78/HSPA5 | |
| Western Blot 1:1000-1:2000, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| 3309 | |
| Full length human GRP78 (Bip) his tagged at the N terminus | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| 78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2 | |
| HSPA5 | |
| IgG | |
| Protein A purified | |
| Detects a protein with a mass of approx. 78kDa corresponding to Grp78. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title