Learn More
GRP94 Mouse anti-Human, Unconjugated, Clone: 4G7C7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 60012-2-IG-150UL
 View more versions of this product
Product Code. 16806975
  • € 410.00 / 150µL
Description
The GRP94 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GRP94. This antibody recognizes human antigen. The GRP94 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
Specifications
GRP94
Monoclonal
853 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry
4G7C7
Unconjugated
150 μL
7184