missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GRP94 Recombinant Rabbit Monoclonal Antibody (7P4V7)

Product Code. p-200111374
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30282785 20 μL 20µL
30284025 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30282785 Supplier Invitrogen™ Supplier No. MA557945

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: EPLLNWMKDK ALKDKIEKAV VSQRLTESPC ALVASQYGWS GNMERIMKAQ AYQTGKDIST NYYASQKKTF EINPRHPLIR DMLRRIKEDE DDKTVLDLAV.

GRP94 is a 803 amino acid protein belonging to the heat shock protein 90 family. It acts as a molecular chaperone that functions in the processing and transport of secreted proteins. GRP94 and its N-terminal fragment stimulates CTL expansion and maturation of human monocyte-derived dendritic cells (MDDC). It plays a central role in innate as well as acquired immunity, maturation and chemotaxis of dendritic cells, Ab production, cross-priming, as a potential marker in breast cancer and is a peptide acceptor in endoplasmic reticulum and an accessory to peptide loading of MHC class I molecules. Expression of GRP94 suppressed A23187-induced apoptosis and stabilized calcium homeostasis. GRP94 is expressed in melanoma or liver metastases of colon carcinoma cells, human gastric carcinoma BGC-823 cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRP94
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 7P4V7
Concentration 1.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene HSP90B1
Gene Accession No. P08113, P14625, Q66HD0
Gene Alias 108K heat shock protein; 108K HSP; 94 kDa glucose-regulated protein; 94 kDa glucose-regulated protein {ECO:0000250; 98 kDa protein kinase; ecgp; endoplasmic reticulum resident protein 99; Endoplasmin; endoplasmin {ECO:0000250; endoplasmin-like protein; endothelial cell (HBMEC) glycoprotein; epididymis luminal protein 35; epididymis secretory sperm binding protein Li 125m; ERp99; glucose-regulated protein; glucose-regulated protein GRP94; glucose-regulated protein GRP94 precursor; GP96; gp96 homolog; gp96/GRP94; GRP94; GRP-94; heat shock 108 kDa protein; heat shock protein 108; heat shock protein 108.; heat shock protein 90 beta family member 1; heat shock protein 90 kDa beta member 1; heat shock protein 90 kDa beta member 1 {ECO:0000250; heat shock protein 90, beta (Grp94), member 1; heat shock protein 90, beta, member 1; heat shock protein 90kDa beta (Grp94), member 1; heat shock protein 90kDa beta family member 1; heat shock protein 90kDa beta family member 1 L homeolog; heat shock protein 90kDa beta, member 1; heat shock protein gp96; HEL35; HEL-S-125m; HSP 108; hsp108; HSP90 beta; hsp90b1; hsp90b1 {ECO:0000250; hsp90b1.L; Polymorphic tumor rejection antigen 1; PPK 98; Ppk 98; a protein kinase; ppk98; stress-inducible tumor rejection antigen gp96; TA-3; Targ2; tra1; Tra-1; transferrin-binding protein; transforming growth factor alpha regulated gene 2; tumor rejection antigen; tumor rejection antigen (gp96) 1; tumor rejection antigen 1; Tumor rejection antigen gp96; UniProtKB:P08113}; XELAEV_18017384mg
Gene Symbols HSP90B1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 616-715 of human HSP90B1 (NP_003290.1).
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22027, 362862, 7184
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.